accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 3386-33-2

IUPAC Name: 1-chlorooctadecane
Synonyms: 1-chlorooctadecane
Molecular Weight (g/mol): 288.94
Molecular Formula: C18H37Cl
InChi Key: VUQPJRPDRDVQMN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCl
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)