missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctadecane, Certified Reference Material, 10,000 μg/mL in Dichloromethane, MilliporeSigma™ Supelco™

Catalog No. 111017259
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017259 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017259 Supplier Merck Emd Millipore Supplier No. 47584U

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
CAS 3386-33-2
Product Line TraceCERT
Concentration 10,000 μg/mL in dichloromethane
Synonym 1-Chlorooctadecane solution
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC