missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-o-benzoquinone, 98+%

Catalog No. AAL0016914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0016914 25 g
AAL0016906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0016914 Supplier Thermo Scientific Chemicals Supplier No. L0016914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3383-21-9 | C14H20O2 | 220.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3383-21-9
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00001647
InChI Key NOUZOVBGCDDMSX-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione
PubChem CID 76915
IUPAC Name 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione
SMILES CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C

Specifications

Melting Point 113°C to 117°C
Quantity 25 g
Beilstein 2047944
Formula Weight 220.31
Percent Purity ≥98%
Chemical Name or Material 3,5-Di-tert-butyl-o-benzoquinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-189-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only