missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-4-chlorophenylacetic acid, 97%

Catalog No. AAH6014106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6014106 5 g
AAH6014103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6014106 Supplier Thermo Scientific Chemicals Supplier No. H6014106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3381-73-5 | C8H6BrClO2 | 249.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3381-73-5
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.49
MDL Number MFCD08276760
InChI Key KKOAAWLOOHBFQP-UHFFFAOYNA-N
Synonym 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid
PubChem CID 10490868
IUPAC Name 2-bromo-2-(4-chlorophenyl)acetic acid
SMILES OC(=O)C(Br)C1=CC=C(Cl)C=C1

Specifications

Melting Point 95°C to 99°C
Quantity 5 g
UN Number UN3261
Formula Weight 249.49
Percent Purity 97%
Chemical Name or Material alpha-Bromo-4-chlorophenylacetic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only