missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-4-chlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. B33675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33675G 5 g
B336725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33675G Supplier TCI America Supplier No. B33675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3381-73-5
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.49
MDL Number MFCD08276760
InChI Key KKOAAWLOOHBFQP-UHFFFAOYNA-N
Synonym 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid
PubChem CID 10490868
IUPAC Name 2-bromo-2-(4-chlorophenyl)acetic acid
SMILES OC(=O)C(Br)C1=CC=C(Cl)C=C1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3261
Formula Weight 249.49
Percent Purity ≥98.0% (T)
Chemical Name or Material alpha-Bromo-4-chlorophenylacetic Acid