accessibility menu, dialog, popup

UserName

H3CBrCH3

CAS RN 3377-87-5

IUPAC Name: 3-bromohexane
Synonyms: hexane, 3-bromo-
Molecular Weight (g/mol): 165.07
Molecular Formula: C6H13Br
InChi Key: IOZOJWNUKLCDML-UHFFFAOYNA-N
SMILES: CCCC(Br)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results