accessibility menu, dialog, popup

UserName

CAS RN 3373-53-3

IUPAC Name: 6-amino-2,7-dihydro-1H-purin-2-one
Synonyms: 6-amino-1,7-dihydropurin-2-one
Molecular Weight (g/mol): 151.13
Molecular Formula: C5H5N5O
InChi Key: DRAVOWXCEBXPTN-UHFFFAOYSA-N
SMILES: NC1=C2NC=NC2=NC(=O)N1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)