missing translation for 'onlineSavingsMsg'
Learn More

Isoguanine 98.0+%, TCI America™

Catalog No. I0370100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0370100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0370100MG Supplier TCI America Supplier No. I0370100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3373-53-3
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00142931
InChI Key DRAVOWXCEBXPTN-UHFFFAOYSA-N
Synonym isoguanine, 2-hydroxyadenine, 2-hydroxy-6-aminopurine, 6-amino-3,7-dihydro-2h-purin-2-one, 6-amino-2-hydroxypurine, 6-amino-7h-purin-2-ol, 2-oxoadenine, 6-amino-1,3-dihydro-2h-purin-2-one, 2-oxyadenine, 6-amino-9h-purin-2-ol
PubChem CID 76900
ChEBI CHEBI:62462
IUPAC Name 6-amino-2,7-dihydro-1H-purin-2-one
SMILES NC1=C2NC=NC2=NC(=O)N1

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 151.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Isoguanine