missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-aminopurine, 98%

Catalog No. AC241752500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241752500 250 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC241752500 Supplier Thermo Scientific Chemicals Supplier No. 241752500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3373-53-3 | C5H5N5O | 151.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3373-53-3
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00142931
InChI Key DRAVOWXCEBXPTN-UHFFFAOYSA-N
Synonym isoguanine, 2-hydroxyadenine, 2-hydroxy-6-aminopurine, 6-amino-3,7-dihydro-2h-purin-2-one, 6-amino-2-hydroxypurine, 6-amino-7h-purin-2-ol, 2-oxoadenine, 6-amino-1,3-dihydro-2h-purin-2-one, 2-oxyadenine, 6-amino-9h-purin-2-ol
PubChem CID 76900
ChEBI CHEBI:62462
IUPAC Name 6-amino-1,7-dihydropurin-2-one
SMILES NC1=C2NC=NC2=NC(=O)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in alkali,practically insoluble in most organic solvents
Formula Weight 151.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Hydroxy-6-aminopurine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
EINECSNumber 222-157-6

RUO – Research Use Only