accessibility menu, dialog, popup

UserName

HO(R)O(S)OO(R)O(S)(R)HO(R)HOOHO(R)OH(R)OH(S)OH(S)HO(R)OH(S)HO(S)HO

CAS RN 33401-87-5

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(3S,4S,5R,6R)-3,4,5-trihydroxy-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-2-yl]methoxy}oxane-3,4,5-triol
Synonyms: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(3S,4S,5R,6R)-3,4,5-trihydroxy-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-2-yl]methoxy}oxane-3,4,5-triol
Molecular Weight (g/mol): 504.44
Molecular Formula: C18H32O16
InChi Key: OWEGMIWEEQEYGQ-RKEGTIJYSA-N
SMILES: OC[C@H]1O[C@H](OCC2O[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results