missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Panose, ≥97% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 33401-87-5 |
| Linear Formula | C18H32O16 |
| MDL Number | MFCD00151376 |
| Synonym | alpha-D-Glc-(1->6)-alpha-D-Glc-(1->4)-D-Glc; O-alpha-D-Glucopyranosyl-(1->6)-O-alpha-D-glucopyranosyl-(1->4)-D-glucose |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 504.44 |
| Percent Purity | ≥97% (HPLC) |
| Grade | Analytical Standard |