CAS RN 333432-27-2
CAS RN 333432-27-2
IUPAC Name:
4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine
Synonyms:
4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine
Molecular Weight (g/mol):
294.95
Molecular Formula:
C5HBr2N3S
InChi Key:
LEHZIBSAFRVAJP-UHFFFAOYSA-N
SMILES:
BrC1=CN=C(Br)C2=NSN=C12
1
–
1
of
1
results
1
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine 97.0+%, TCI America™
CAS: 333432-27-2 Molecular Formula: C5HBr2N3S Molecular Weight (g/mol): 294.952 MDL Number: MFCD23703147 InChI Key: LEHZIBSAFRVAJP-UHFFFAOYSA-N PubChem CID: 58129789 IUPAC Name: 4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine SMILES: C1=C(C2=NSN=C2C(=N1)Br)Br