CAS RN 333353-44-9
CAS RN 333353-44-9
IUPAC Name:
N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)ethanediamide
Synonyms:
N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)ethanediamide
Molecular Weight (g/mol):
337.85
Molecular Formula:
C17H24ClN3O2
InChi Key:
ZKXLQCIOURANAD-UHFFFAOYSA-N
SMILES:
CC1(C)CC(CC(C)(C)N1)NC(=O)C(=O)NC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
NBD-556 98.0+%, TCI America™
CAS: 333353-44-9 Molecular Formula: C17H24ClN3O2 Molecular Weight (g/mol): 337.848 MDL Number: MFCD00469538 InChI Key: ZKXLQCIOURANAD-UHFFFAOYSA-N Synonym: N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide PubChem CID: 1570601 IUPAC Name: N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)oxamide SMILES: CC1(CC(CC(N1)(C)C)NC(=O)C(=O)NC2=CC=C(C=C2)Cl)C