missing translation for 'onlineSavingsMsg'
Learn More

NBD-556 98.0+%, TCI America™

Catalog No. N109025MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N109025MG 25 mg
N1090100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N109025MG Supplier TCI America Supplier No. N109025MG
Only null left

Chemical Identifiers

CAS 333353-44-9
Molecular Formula C17H24ClN3O2
Molecular Weight (g/mol) 337.848
MDL Number MFCD00469538
InChI Key ZKXLQCIOURANAD-UHFFFAOYSA-N
Synonym N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide
PubChem CID 1570601
IUPAC Name N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)oxamide
SMILES CC1(CC(CC(N1)(C)C)NC(=O)C(=O)NC2=CC=C(C=C2)Cl)C

Specifications

Quantity 25 mg
UN Number 2811
Formula Weight 337.85
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material NBD-556
TSCA No
Recommended Storage Refrigerator