accessibility menu, dialog, popup

UserName

CH3OPSOCH3OCH3NNCH3H3C

CAS RN 333-41-5

IUPAC Name: O,O-diethyl O-6-methyl-2-(propan-2-yl)pyrimidin-4-yl phosphorothioate
Synonyms: diazinon caswell no. 342 nucidol dazzel kleen-dok basudin 10 G dimpylate drawizon exodin disonex
Molecular Weight (g/mol): 304.34
Molecular Formula: C12H21N2O3PS
InChi Key: FHIVAFMUCKRCQO-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CC(C)=NC(=N1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results