accessibility menu, dialog, popup

UserName

CAS RN 333-31-3

IUPAC Name: [2-(acetyloxy)propyl]trimethylazanium bromide
Synonyms: methacholine bromide
Molecular Weight (g/mol): 240.14
Molecular Formula: C8H18BrNO2
InChi Key: MMVPLEUBMWUYIB-UHFFFAOYNA-M
SMILES: [Br-].CC(C[N+](C)(C)C)OC(C)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)