missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-beta-methylcholine bromide, 99%

Catalog No. AC226970250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226970250 25 g Glass bottle
AC226970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226970250 Supplier Thermo Scientific Chemicals Supplier No. 226970250

CAS: 333-31-3 | C8H18BrNO2 | 240.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 333-31-3
Molecular Formula C8H18BrNO2
Molecular Weight (g/mol) 240.14
MDL Number MFCD00011816
InChI Key MMVPLEUBMWUYIB-UHFFFAOYNA-M
Synonym methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide
PubChem CID 92754
IUPAC Name 2-acetyloxypropyl(trimethyl)azanium;bromide
SMILES [Br-].CC(C[N+](C)(C)C)OC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 150°C to 152°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dried substance) (Argentometry)
Packaging Glass bottle
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Br
Quantity 25 g
Beilstein 04, III, 758
Merck Index 10, 5793
Solubility Information Solubility in water: soluble
Formula Weight 240.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetyl-β-methylcholine bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 206-372-2
TSCA TSCA

RUO – Research Use Only