missing translation for 'onlineSavingsMsg'
Learn More

Methacholine Bromide 99.0+%, TCI America™

Catalog No. M007225G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M007225G 25 g
M0072100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M007225G Supplier TCI America Supplier No. M007225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333-31-3
Molecular Formula C8H18BrNO2
Molecular Weight (g/mol) 240.14
MDL Number MFCD00011816
InChI Key MMVPLEUBMWUYIB-UHFFFAOYNA-M
Synonym methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide
PubChem CID 92754
IUPAC Name [2-(acetyloxy)propyl]trimethylazanium bromide
SMILES [Br-].CC(C[N+](C)(C)C)OC(C)=O

Specifications

Melting Point 147°C
Color White
Quantity 25 g
Formula Weight 240.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methacholine Bromide