accessibility menu, dialog, popup

UserName

ClNH3NOO

CAS RN 33240-96-9

IUPAC Name: 3-nitroanilinium chloride
Synonyms: 3-nitroanilinium chloride
Molecular Weight (g/mol): 174.58
Molecular Formula: C6H7ClN2O2
InChi Key: FWUUMDDHHPVSPL-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results