accessibility menu, dialog, popup

UserName

CAS RN 332-77-4

IUPAC Name: 2,5-dimethoxy-2,5-dihydrofuran
Synonyms: 2,5-dimethoxy-2,5-dihydrofuran
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: WXFWXFIWDGJRSC-UHFFFAOYNA-N
SMILES: COC1OC(OC)C=C1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)