accessibility menu, dialog, popup

UserName

CH3NNH3CNS

CAS RN 331717-63-6

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; cyanosulfanide
Synonyms: 1-ethyl-3-methylimidazolium thiocyanate
Molecular Weight (g/mol): 169.25
Molecular Formula: C7H11N3S
InChi Key: VASPYXGQVWPGAB-UHFFFAOYSA-M
SMILES: [S-]C#N.CCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)