missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium thiocyanate, 98%, Thermo Scientific Chemicals

Catalog No. AAH5949306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5949306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5949306 Supplier Thermo Scientific Chemicals Supplier No. H5949306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 331717-63-6 | Molecular Formula: C7H11N3S | Molecular Weight: 169.246 g/mol

It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 331717-63-6
Molecular Formula C7H11N3S
Molecular Weight (g/mol) 169.246
MDL Number MFCD06798182
InChI Key VASPYXGQVWPGAB-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium thiocyanate, basionics™ vs 01, dsstox_cid_27860, dsstox_rid_82613, dsstox_gsid_47884, 1-methyl-3-ethylimidazolium thiocyanate, 1-ethyl-3-methyl imidazolium thio-cyanate, 1-ethyl-3-methyl-1h-imidazol-3-ium thiocyanate, 1-ethyl-3-methylimidazolium thiocyanate h-nmr, hplc, 1-ethyl-3-methylimidazolium thiocyanate, for electrochemistry t
PubChem CID 16211115
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;thiocyanate
SMILES CCN1C=C[N+](=C1)C.C(#N)[S-]

Specifications

Melting Point -6°C
Density 1.14
Refractive Index n20/D 1.553
Quantity 5 g
UN Number UN2810
Sensitivity Moisture sensitive
Formula Weight 169.25
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium thiocyanate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only