accessibility menu, dialog, popup

UserName

CAS RN 3303-84-2

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]propanoic acid|BOC-BETA-ALA-OH
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: WCFJUSRQHZPVKY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCC(O)=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)