accessibility menu, dialog, popup

UserName

CAS RN 328090-25-1

CH3NNH3CCH3SOOO

CAS RN 328090-25-1

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium 4-methylbenzene-1-sulfonate
Synonyms: 1-ethyl-3-methylimidazolium tosylate
Molecular Weight (g/mol): 282.36
Molecular Formula: C13H18N2O3S
InChi Key: HXMUPILCYSJMLQ-UHFFFAOYSA-M
SMILES: CCN1C=C[N+](C)=C1.CC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results