missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. E06515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06515G 5 g
E065125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06515G Supplier TCI America Supplier No. E06515G
Only null left

Chemical Identifiers

CAS 328090-25-1
Molecular Formula C13H18N2O3S
Molecular Weight (g/mol) 282.358
InChI Key HXMUPILCYSJMLQ-UHFFFAOYSA-M
PubChem CID 10636633
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;4-methylbenzenesulfonate
SMILES CCN1C=C[N+](=C1)C.CC1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

Quantity 5 g
Formula Weight 282.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium p-Toluenesulfonate