accessibility menu, dialog, popup

UserName

H3CNCH3H3COHSOOOCH3

CAS RN 32503-27-8

IUPAC Name: tetrabutylazanium hydrogen sulfate
Synonyms: tetrabutylammonium ion hydrogen sulfate
Molecular Weight (g/mol): 339.54
Molecular Formula: C16H37NO4S
InChi Key: SHFJWMWCIHQNCP-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (5)
  • (3)
  • (3)
  • (4)
  • (6)
Quantity 
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (5)
  • (4)
  • (3)
  • (2)
  • (4)
Grade 
  • (4)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (3)
  • (5)
  • (13)
  • (5)
  • (3)
  • (3)
  • (4)
  • (6)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (4)
  • (4)
  • (2)
  • (3)