accessibility menu, dialog, popup

UserName

CAS RN 3236-82-6

IUPAC Name: pentaethoxyniobium
Synonyms: niobium(V) ethoxide
Molecular Weight (g/mol): 318.21
Molecular Formula: C10H25NbO5
InChi Key: ZTILUDNICMILKJ-UHFFFAOYSA-N
SMILES: CCO[Nb](OCC)(OCC)(OCC)OCC
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)