missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) ethoxide, 99.9% (metals basis)

Catalog No. AAL1603106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1603106 5 g
AAL1603103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1603106 Supplier Thermo Scientific Chemicals Supplier No. L1603106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3236-82-6 | C10H25NbO5 | 318.21 g/mol

Niobium(V) ethoxide is used in nano-materials & thin films, catalysis & synthesis, hybrid & disperse materials, ferroelectric metals. It is mainly used for the sol-gel processing of materials containing niobium oxides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Niobium(V) ethoxide is used in nano-materials & thin films, catalysis & synthesis, hybrid & disperse materials, ferroelectric metals. It is mainly used for the sol-gel processing of materials containing niobium oxides

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. It is sensitive to moisture. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 3236-82-6
Molecular Formula C10H25NbO5
Molecular Weight (g/mol) 318.21
MDL Number MFCD00015122
InChI Key ZTILUDNICMILKJ-UHFFFAOYSA-N
Synonym niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
PubChem CID 160675
SMILES CCO[Nb](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 140°C to 142°C (0.1mmHg)
Melting Point 6°C
Quantity 5 g
UN Number UN2924
Beilstein 3678883
Flash Point 36°C (97°F)
Solubility Information Reacts with water.
IUPAC Name ethanolate;niobium(5+)
Formula Weight 318.21
Percent Purity 99.9%
Refractive Index 1.516
Sensitivity Moisture sensitive
Density 1.258
Assay (metals basis)
Chemical Name or Material Niobium(V) ethoxide
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 221-795-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only