missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) ethoxide, 99.9%, (trace metal basis)

Catalog No. AC316880050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC316880050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC316880050 Supplier Thermo Scientific Chemicals Supplier No. 316880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3236-82-6 | C10H25NbO5 | 318.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 3236-82-6
Molecular Formula C10H25NbO5
Molecular Weight (g/mol) 318.21
MDL Number MFCD00015122
InChI Key ZTILUDNICMILKJ-UHFFFAOYSA-N
Synonym niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
PubChem CID 160675
SMILES CCO[Nb](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 140.0°C to 142.0°C (0.1mmHg)
Melting Point 6.0°C
CAS Min % 99.9
CAS Max % 100.0
Physical Form Liquid
Quantity 5 g
Assay Percent Range 99.9%
Linear Formula Nb(OC2H5)5
Flash Point 36°C
Solubility Information Solubility in water: reacts
IUPAC Name pentaethoxyniobium
Formula Weight 318.21
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 1.268
Density 1.2680g/mL
Chemical Name or Material Niobium(V) ethoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 221-795-2

RUO – Research Use Only