accessibility menu, dialog, popup

UserName

ClH3COONH2OOCH3

CAS RN 32213-95-9

IUPAC Name: 1,4-dimethyl 2-aminobutanedioate hydrochloridyl
Synonyms: 1,4-dimethyl 2-aminobutanedioate hydrochloridyl
Molecular Weight (g/mol): 196.61
Molecular Formula: C6H11ClNO4
InChi Key: KUSLQZQCHPWKKH-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)CC(N)C(=O)OC
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)