accessibility menu, dialog, popup

UserName

CAS RN 321-30-2

IUPAC Name: bis(7H-purin-6-amine); sulfuric acid
Synonyms: adenine sulfate
Molecular Weight (g/mol): 368.33
Molecular Formula: C10H12N10O4S
InChi Key: LQXHSCOPYJCOMD-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)