missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine sulfate, 98+%

Catalog No. AAA1696418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1696418 50 g
AAA1696409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1696418 Supplier Thermo Scientific Chemicals Supplier No. A1696418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol

It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.

Solubility
Soluble in water (4 mg/ml), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and HCl (10 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name 7H-purin-6-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Melting Point ∼210°C (decomposition)
Odor Odorless
Assay Percent Range >98%
Quantity 50 g
Beilstein 3820263
Sensitivity Hygroscopic
Merck Index 14,152
Solubility Information Soluble in water (4mg/ml),ethanol (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),and HCl (10mg/ml).
Formula Weight 368.34
Percent Purity ≥98%
Chemical Name or Material Adenine sulfate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-286-5
RTECSNumber AU7140000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only