missing translation for 'onlineSavingsMsg'
Learn More

Adenine Sulfate, MP Biomedicals

Catalog No. MP114014522
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114014522 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114014522 Supplier MP Biomedicals Inc Supplier No. 114014522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name bis(7H-purin-6-amine); sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Quantity 10 g
Physical Form Powder
Chemical Name or Material ADENINE SULFATE
Recommended Storage Store at Room Temperature.