accessibility menu, dialog, popup

UserName

HOONOO

CAS RN 3130-75-4

IUPAC Name: 4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)butanoic acid
Synonyms: 4-phthalimidobutyric acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C12H11NO4
InChi Key: HMKSXJBFBVGLJJ-UHFFFAOYSA-N
SMILES: OC(=O)CCCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results