missing translation for 'onlineSavingsMsg'
Learn More

4-Phthalimidobutyric Acid 98.0+%, TCI America™

Catalog No. P21121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21121G Supplier TCI America Supplier No. P21121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3130-75-4
Molecular Formula C12H11NO4
Molecular Weight (g/mol) 233.223
MDL Number MFCD00196079
InChI Key HMKSXJBFBVGLJJ-UHFFFAOYSA-N
PubChem CID 18411
IUPAC Name 4-(1,3-dioxoisoindol-2-yl)butanoic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCC(=O)O

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Formula Weight 233.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phthalimidobutyric Acid
RTECSNumber NR3459000
TSCA No