accessibility menu, dialog, popup

UserName

CAS RN 309735-86-2

H3CPCH3PH3CCH3H3CPCH3PH3CCH3H3CNH2CH3ClClClClClRuRu(?)(?)

CAS RN 309735-86-2

IUPAC Name: bis(λ²-ruthenium(2+)) dimethylazanium bis({2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane) pentachloride
Synonyms: bis(λ²-ruthenium(2+)) dimethylazanium bis({2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane) pentachloride
Molecular Weight (g/mol): 1783.07
Molecular Formula: C98H88Cl5NP4Ru2
InChi Key: FAAFCSOHMVBJIS-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].C[NH2+]C.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results