missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[NH2Me2][(RuCl((S)-tolbinap))2(mu-Cl)3], TCI America™

$71.81 - $249.03

Chemical Identifiers

CAS 309735-86-2
Molecular Formula C98H88Cl5NP4Ru2
Molecular Weight (g/mol) 1783.074
MDL Number MFCD09753034
InChI Key FAAFCSOHMVBJIS-UHFFFAOYSA-J
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
PubChem CID 131675153
IUPAC Name [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.Cl.[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N1018200MG View Documents TCI America
N1018200MG
200mg Each for $71.81
 
N10181G View Documents TCI America
N10181G
1g Each for $249.03
 
Specifications

Chemical Identifiers

309735-86-2
1783.074
FAAFCSOHMVBJIS-UHFFFAOYSA-J
131675153
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.Cl.[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2]
C98H88Cl5NP4Ru2
MFCD09753034
Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride

Specifications

Undesignated
309735-86-2
MFCD09753034
FAAFCSOHMVBJIS-UHFFFAOYSA-J
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride
131675153
Crystal-Powder at 20°C
200mg
C98H88Cl5NP4Ru2
Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.Cl.[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2]
1783.074
1783.07
[NH2Me2][(RuCl((S)-tolbinap))2(mu-Cl)3]
Safety and Handling

Safety and Handling

TSCA : No