CAS RN 30968-45-7
IUPAC Name:
methyl (1R,4aR,4bS,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,8a,9,10,10a-dodecahydrophenanthrene-1-carboxylate
Synonyms:
methyl (1R,4aR,4bS,10aR)-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,8a,9,10,10a-decahydrophenanthrene-1-carboxylate
Molecular Weight (g/mol):
318.5
Molecular Formula:
C21H34O2
InChi Key:
MARRJGBPDCCAEK-HSLMUJISSA-N
SMILES:
COC(=O)[C@]1(C)CCC[C@]2(C)[C@H]3CCC(=CC3CC[C@@H]12)C(C)C
1
–
1
of
1
results
1
Methyl Dihydroabietate, TCI America™
CAS: 30968-45-7 Molecular Formula: C21H34O2 MDL Number: MFCD00466887 Synonym: Dihydroabietic Acid Methyl Ester