missing translation for 'onlineSavingsMsg'
Learn More

Methyl Dihydroabietate, TCI America™

Catalog No. D108825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D108825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D108825G Supplier TCI America Supplier No. D108825G
Only null left

Chemical Identifiers

CAS 30968-45-7
Molecular Formula C21H34O2
MDL Number MFCD00466887
Synonym Dihydroabietic Acid Methyl Ester

Specifications

Boiling Point 365°C
Quantity 25 g
Formula Weight 318.50
Physical Form Liquid
Chemical Name or Material Methyl Dihydroabietate