accessibility menu, dialog, popup

UserName

CH3OHH3CH2N

CAS RN 3096-70-6

IUPAC Name: 4-amino-3,5-dimethylphenol
Synonyms: 4-hydroxy-2,6-dimethylaniline 1,3-dimethyl-2-amino-5-xylenol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: GCWYXRHXGLFVFE-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results