accessibility menu, dialog, popup

UserName

ONOO

CAS RN 3096-52-4

IUPAC Name: 2-nitro-9H-fluoren-9-one
Synonyms: 2-nitrofluoren-9-one
Molecular Weight (g/mol): 225.2
Molecular Formula: C13H7NO3
InChi Key: AJEAHBZZHSLIQP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results