accessibility menu, dialog, popup

UserName

CH3NHO

CAS RN 30727-14-1

IUPAC Name: 1-ethylpyrrolidin-3-ol
Synonyms: 3-pyrrolidinol, 1-ethyl-
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: IDBNECMSCRAUNU-UHFFFAOYNA-N
SMILES: CCN1CCC(O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results