missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-pyrrolidinol 95.0+%, TCI America™

Catalog No. E042510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E042510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E042510ML Supplier TCI America Supplier No. E042510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30727-14-1
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003179
InChI Key IDBNECMSCRAUNU-UHFFFAOYNA-N
Synonym 1-Ethyl-3-hydroxypyrrolidine
PubChem CID 98203
IUPAC Name 1-ethylpyrrolidin-3-ol
SMILES CCN1CCC(O)C1

Specifications

Boiling Point 110°C
Quantity 10 mL
Formula Weight 115.18
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-pyrrolidinol
RTECSNumber UY5685500
TSCA No