accessibility menu, dialog, popup

UserName

CAS RN 3066-71-5

IUPAC Name: cyclohexyl prop-2-enoate
Synonyms: cyclohexyl acrylate
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: KBLWLMPSVYBVDK-UHFFFAOYSA-N
SMILES: C=CC(=O)OC1CCCCC1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)