accessibility menu, dialog, popup

UserName

CAS RN 30562-34-6

IUPAC Name: 13-hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
Synonyms: geldanamycin
Molecular Weight (g/mol): 560.64
Molecular Formula: C29H40N2O9
InChi Key: QTQAWLPCGQOSGP-UHFFFAOYNA-N
SMILES: COC1CC(C)CC2=C(OC)C(=O)C=C(NC(=O)C(C)=CC=CC(OC)C(OC(N)=O)C(C)=CC(C)C1O)C2=O
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)