accessibility menu, dialog, popup

UserName

H3COOOOOOOOOH

CAS RN 3055-98-9

IUPAC Name: 3,6,9,12,15,18,21,24-octaoxahexatriacontan-1-ol
Synonyms: dodecyloctaethyleneglycol monoether octaethyleneglycol monododecyl ether
Molecular Weight (g/mol): 538.76
Molecular Formula: C28H58O9
InChi Key: YYELLDKEOUKVIQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results