missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ C12E8 Detergent, Protein Grade™, 10% Solution,Calbiochem™,

Catalog No. 2055321SET
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2055321SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2055321SET Supplier MilliporeSigma™ Supplier No. 2055321SET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3055-98-9
Molecular Formula C28H58O9
Molecular Weight (g/mol) 538.763
InChI Key YYELLDKEOUKVIQ-UHFFFAOYSA-N
PubChem CID 123921
ChEBI CHEBI:41527
IUPAC Name 2-[2-[2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Quantity 1 Set
Packaging Glass Bottle
Grade Protein