accessibility menu, dialog, popup

UserName

NH2SOHO

CAS RN 3054-01-1

IUPAC Name: 2-amino-3-(benzylsulfanyl)propanoic acid
Synonyms: benzylcysteine
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: GHBAYRBVXCRIHT-UHFFFAOYNA-N
SMILES: NC(CSCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)