missing translation for 'onlineSavingsMsg'
Learn More

S-Benzyl-L-cysteine 98.0+%, TCI America™

Catalog No. B08655G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B08655G Supplier TCI America Supplier No. B08655G

Specifications

CAS 3054-01-1
Color White
Molecular Formula C10H13NO2S
Quantity 5 g
MDL Number MFCD00002613
Synonym s-benzyl-l-cysteine, benzylcysteine, h-cys bzl-oh, s-benzylcysteine, poly-s-benzylcysteine, r-2-amino-3-benzylthio propanoic acid, cysteine, s-phenylmethyl, chembl63130, unii-9vre13m548, 3-benzylthioalanine
InChI Key GHBAYRBVXCRIHT-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)CSCC(C(=O)O)N
IUPAC Name (2R)-2-amino-3-benzylsulfanylpropanoic acid
Molecular Weight (g/mol) 211.28
PubChem CID 193613
ChEBI CHEBI:41085
Formula Weight 211.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material S-Benzyl-L-cysteine
Show More Show Less
EINECSNumber (9)-1591
TSCA Yes