missing translation for 'onlineSavingsMsg'
Learn More

S-Benzyl-L-cysteine, 99%, Thermo Scientific Chemicals

Catalog No. AAL1166614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1166614 25 g
AAL1166606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1166614 Supplier Thermo Scientific Chemicals Supplier No. L1166614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3054-01-1
MDL Number MFCD00002613

Specifications

Melting Point ∽214°C (decomposition)
Molecular Formula C7H12O4
Beilstein 1879358
Synonym 3-methyladipic acid, hexanedioic acid, 3-methyl, +-3-methyladipic acid, beta-methyl-adipic acid, 3-methyl-hexanedioic acid, 3-methyl adipic acid, 3-methyl-adipic acid, acmc-20ah5v, acmc-1cr8a, hexanedioic acid,3-methyl
InChI Key SYEOWUNSTUDKGM-UHFFFAOYNA-N
SMILES CC(CCC(=O)O)CC(=O)O
IUPAC Name 3-methylhexanedioic acid
Quantity 25 g
Molecular Weight (g/mol) 160.17
PubChem CID 12292
ChEBI CHEBI:68503
Formula Weight 211.28
Optical Rotation +28° (c=1.5 in 1N NaOH)
Percent Purity 99%
Chemical Name or Material S-Benzyl-L-cysteine
Show More Show Less
EINECSNumber 221-273-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only